2-(3,5-Dimethylphenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

043252-500MG 043252-1G 043252-5G
101859-900EA 556.92 USD
101859-900 101859-902 101859-904
2-(3,5-Dimethylphenoxy)benzaldehyde ≥95%
2-(3,5-Dimethylphenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₂
Boiling Pt: 122-124°C/0.05mm
MDL Number: MFCD01568762
CAS Number: 320423-51-6

Order Now

Specification Test Results

Boiling point 122-124°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR