2-Methyl-2-(phenylsulfanyl)propanal ≥95%

Supplier: Matrix Scientific

043227-500MG 043227-1G 043227-5G
101859-782EA 375.38 USD
101859-782 101859-784 101859-786
2-Methyl-2-(phenylsulfanyl)propanal ≥95%
2-Methyl-2-(phenylsulfanyl)propanal

Hazard: Irritant

Formula: C₁₀H₁₂OS
MW: 180.27 g/mol
Boiling Pt: 112-116°C/4mm
MDL Number: MFCD00243420
CAS Number: 63996-66-7

Order Now

Specification Test Results

Boiling point 112-116°C/4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR