4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043172-500MG 043172-1G 043172-5G
101859-494EA 195.38 USD
101859-494 101859-496 101859-498
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₇ClF₃NO₂
MW: 301.65 g/mol
Boiling Pt: 106°C/0.1mm
MDL Number: MFCD09607890
CAS Number: 950994-19-1

Order Now

Specification Test Results

Boiling point 106°C/0.1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR