Ethyl-3-(2-furyl)-3-oxopropanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(2-Furyl)-3-oxopropanoate

043145-500MG 043145-1G 043145-5G
101859-374EA 375.38 USD
101859-374 101859-376 101859-378
Ethyl-3-(2-furyl)-3-oxopropanoate ≥95%
Ethyl-3-(2-furyl)-3-oxopropanoate

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 114 °C (2 mmHg)
MDL Number: MFCD00122473
CAS Number: 615-09-8

Order Now

Specification Test Results

Boiling point 114°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR