1-(1,3-Thiazol-2-yl)-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(Thiazol-2-yl)-1H-pyrrole-2-carbaldehyde
043063-500MG
043063-5G
101858-958EA
136.92
USD
101858-958
101858-962
1-(1,3-Thiazol-2-yl)-1H-pyrrole-2-carbaldehyde ≥95%
1-(1,3-Thiazol-2-yl)-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₈H₆N₂OS MW: 178.21 g/mol Boiling Pt: 135 °C (2 mmHg) |
MDL Number:
MFCD02665165 CAS Number: 383136-31-0 |
Specification Test Results
Boiling point | 135°C/2mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...