2-(N-Methyl-2-pyridylamino)ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Methyl(pyridin-2-yl)amino)ethanol, N-Methyl-N-(2-pyridyl)ethanolamine, 2-[N-(2-Hydroxyethyl)-N-methylamino]pyridine

043009-500MG 043009-5G
101858-694EA 24.62 USD
101858-694 101858-698
2-(N-Methyl-2-pyridylamino)ethanol ≥95%
2-(N-Methyl-2-pyridylamino)ethanol

Hazard: Irritant

Formula: C₈H₁₂N₂O
MW: 152.20 g/mol
Boiling Pt: 98…100 °C (0.1 mmHg)
MDL Number: MFCD02667633
CAS Number: 122321-04-4

Order Now

Specification Test Results

Boiling point 98-100°C/0.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR