Methyl-2-(2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
042869-500MG
042869-5G
101857-958EA
278.46
USD
101857-958
101857-962
Methyl-2-(2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate ≥95%
Methyl-2-(2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₅NO₂ Boiling Pt: 86°C/0.1mm |
MDL Number:
MFCD01125269 CAS Number: 83935-44-8 |
Specification Test Results
Boiling point | 86°C/0.1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...