Methyl-2-formylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-formylbenzoate

042839-1G 042839-5G
101857-796EA 29.23 USD
101857-796 101857-798
Methyl-2-formylbenzoate ≥95%
Methyl-2-formylbenzoate

Hazard: Irritant/Refrigerate

Formula: C₉H₈O₃
MW: 164.1580 g/mol
Boiling Pt: 76°C/0.1mm
MDL Number: MFCD00204231
CAS Number: 4122-56-9

Order Now

Specification Test Results

Boiling point 76°C/0.1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR