5-(Chloromethyl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole ≥95%

Supplier: Matrix Scientific

042815-500MG 042815-1G 042815-5G
101857-686EA 556.92 USD
101857-686 101857-688 101857-690
5-(Chloromethyl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole ≥95%
5-(Chloromethyl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂O
Boiling Pt: 87-90°C/0.2mm
MDL Number: MFCD06200906
CAS Number: 844498-80-2

Order Now

Specification Test Results

Boiling point 87-90°C/0.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR