Methyl-2-(2-propynyloxy)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
042742-500MG
042742-1G
042742-5G
101857-284EA
461.54
USD
101857-284
101857-286
101857-288
Methyl-2-(2-propynyloxy)benzenecarboxylate ≥95%
Methyl-2-(2-propynyloxy)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀O₃ Boiling Pt: 100°C/0.3mm |
MDL Number:
MFCD06797370 CAS Number: 59155-84-9 |
Specification Test Results
Boiling point | 100°C/0.3mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...