1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-methylpiperazine ≥95%

Supplier: Matrix Scientific

042707-500MG 042707-1G 042707-5G
101857-118EA 278.46 USD
101857-118 101857-120 101857-122
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-methylpiperazine ≥95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-methylpiperazine

Hazard: Irritant

Formula: C₉H₁₄ClN₃S
Boiling Pt: 86 °C (0.1 mmHg)
MDL Number: MFCD01568175
CAS Number: 341967-70-2

Order Now

Specification Test Results

Boiling point 86°C/0.1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR