3-(1-Piperazino)propionitrile ≥97%

Supplier: Matrix Scientific

034753-1G 034753-5G
101838-294EA 32.31 USD
101838-294 101838-296
3-(1-Piperazino)propionitrile ≥97%
3-(1-Piperazino)propionitrile

Hazard: Irritant

Formula: C₇H₁₃N₃
MW: 139.2 g/mol
Boiling Pt: 95-98°C/3mm
MDL Number: MFCD02093566
CAS Number: 34064-86-3

Order Now

Specification Test Results

Boiling point 95-98°C/3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR