3-Chloro-5-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

026741-1G 026741-5G
101822-790EA 84.62 USD
101822-790 101822-792
3-Chloro-5-(trifluoromethyl)pyridine ≥98%
3-Chloro-5-(trifluoromethyl)pyridine

Hazard: Toxic

Formula: C6H3ClF3N
MW: 181.55 g/mol
Boiling Pt: 144 °C
MDL Number: MFCD00042227
CAS Number: 85148-26-1

Order Now

Specification Test Results

Boiling point 144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR