1-(4-Chlorophenyl)-N-methylmethanamine

Supplier: Matrix Scientific

Synonyms: 4-Chloro-N-methylbenzylamine

020395-1G 020395-5G
101809-948EA 33.85 USD
101809-948 101809-950
1-(4-Chlorophenyl)-N-methylmethanamine
1-(4-Chlorophenyl)-N-methylmethanamine

Hazard: Irritant

Formula: C₈H₁₀ClN
MW: 155.6247 g/mol
Boiling Pt: 77 °C
MDL Number: MFCD00018749
CAS Number: 104-11-0

Order Now

Specification Test Results

Boiling point 77 °CC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR