Trifluoroacetaldehyde dimethyl acetal ≥97%

Supplier: Matrix Scientific

012893-1G 012893-5G
101803-576EA 92.31 USD
101803-576 101803-578
Trifluoroacetaldehyde dimethyl acetal ≥97%

Hazard: Flammable

Formula: C₄H₇F₃O₂
Boiling Pt: 78 °C
Density: 1.2
Flash Pt: 4 °C
MDL Number: MFCD00192521
CAS Number: 42415-20-3

Order Now

Specification Test Results

Boiling point 78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR