N-Methyl-N-methoxydifluoroacetamide ≥97%

Supplier: Matrix Scientific

012611-1G 012611-5G
101802-974EA 78.46 USD
101802-974 101802-976
N-Methyl-N-methoxydifluoroacetamide ≥97%
N-Methyl-N-methoxydifluoroacetamide

Hazard: Toxic

Formula: C₄H₇F₂NO₂
Boiling Pt: 40 °C (0.7 mmHg)
MDL Number: MFCD06248152
CAS Number: 142492-01-1

Order Now

Specification Test Results

Boiling point 40°C/0.7mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR