Methyl(4-aminophenyl)acetate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl-(4-aminophenyl)acetate, Methyl 2-(4-aminophenyl)acetate

012593-5G 012593-25G 012593-100G
101802-938EA 36.92 USD
101802-938 101802-940 102832-846
Methyl(4-aminophenyl)acetate ≥98%
Methyl(4-aminophenyl)acetate

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 96…100 °C (0.1 mmHg)
MDL Number: MFCD06200783
CAS Number: 39552-81-3

Order Now

Specification Test Results

Boiling point 96-100°C/0.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR