1-(2-Aminoethyl)-2-imidazolidone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2-Aminoethyl)imidazolidin-2-one

012557-1G 012557-5G
101802-852EA 60 USD
101802-852 101802-854
1-(2-Aminoethyl)-2-imidazolidone ≥98%
1-(2-Aminoethyl)-2-imidazolidone

TCA listed: T
Hazard: Corrosive/Stored under Argon

Formula: C₅H₁₁N₃O
MW: 129.16 g/mol
Boiling Pt: 138…140 °C (0.15 mmHg)
MDL Number: MFCD00086348
CAS Number: 6281-42-1

Order Now

Specification Test Results

Boiling point 138-140°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR