1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥97%

Supplier: Matrix Scientific

007662-1G 007662-5G
101793-198EA 106.15 USD
101793-198 101793-200
1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥97%
1-(Trifluoromethyl)cyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 76°C/14mm
MDL Number: MFCD03425197
CAS Number: 277756-45-3

Order Now

Specification Test Results

Boiling point 76°C/14mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR