1-(4-Morpholino)cyclododecene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(1-Cyclododecen-1-yl)morpholine, 1-Morpholino-1-cyclododecene

007630-5G 007630-25G
101793-136EA 44.62 USD
101793-136 101793-138
1-(4-Morpholino)cyclododecene ≥97%
1-(4-Morpholino)cyclododecene

Hazard: Irritant

Formula: C₁₆H₂₉NO
MW: 251.41 g/mol
Boiling Pt: 180 °C ( 5 mmHg)
MDL Number: MFCD00042788
CAS Number: 3725-39-1

Order Now

Specification Test Results

Boiling point 180°C/ 5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR