3-(4'-Chlorophenyl)propanol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)propan-1-ol

007542-5G 007542-25G
101793-004EA 90.77 USD
101793-004 101793-006
3-(4'-Chlorophenyl)propanol ≥97%
3-(4'-Chlorophenyl)propanol

Hazard: Irritant

Formula: C₉H₁₁ClO
MW: 170.64 g/mol
Boiling Pt: 140 °C ( 10 mmHg)
MDL Number: MFCD00831102
CAS Number: 6282-88-8

Order Now

Specification Test Results

Boiling point 140°C/ 10mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR