1-(4-Chlorophenyl)ethanamine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)ethylamine

007538-1G 007538-5G
101792-994EA 16.92 USD
101792-994 101792-996
1-(4-Chlorophenyl)ethanamine ≥97%
1-(4-Chlorophenyl)ethanamine

Hazard: Irritant

Formula: C₈H₁₀ClN
Boiling Pt: 119°C/20mm
Melting Pt: 140…141 °C
MDL Number: MFCD00044766
CAS Number: 6299-02-1

Order Now

Specification Test Results

Boiling point 119°C/20mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR