2-Chloro-5-methylbenzonitrile ≥97%

Supplier: Matrix Scientific

007533-1G 007533-5G
101792-990EA 63.08 USD
101792-990 101792-992
2-Chloro-5-methylbenzonitrile ≥97%
2-Chloro-5-methylbenzonitrile

Hazard: Irritant-Harmful

Formula: C₈H₆ClN
Boiling Pt: 113 °C (7 mmHg)
MDL Number: MFCD00045595
CAS Number: 4387-32-0

Order Now

Specification Test Results

Boiling point 113°C/7mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR