2-Chloro-N,N-dimethylaniline ≥97%

Supplier: Matrix Scientific

007531-5G 007531-25G
101792-984EA 60 USD
101792-984 101792-986
2-Chloro-N,N-dimethylaniline ≥97%
2-Chloro-N,N-dimethylaniline

Hazard: Irritant-Harmful

Formula: C₈H₁₀ClN
Boiling Pt: 70 °C (7 mmHg)
MDL Number: MFCD03452792
CAS Number: 698-01-1

Order Now

Specification Test Results

Boiling point 70°C/7mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR