2-Bromo-p-toluonitrile ≥97%

Supplier: Matrix Scientific

007455-5G 007455-25G
101792-812EA 24.62 USD
101792-812 101792-814
2-Bromo-p-toluonitrile ≥97%
2-Bromo-p-toluonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₈H₆BrN
Boiling Pt: 110 °C (5 mmHg)
MDL Number: MFCD00158968
CAS Number: 42872-73-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 110°C/5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR