2-Chloro-2,2-difluoroethanol ≥96%

Supplier: Matrix Scientific

007429-5G 007429-25G
101792-742EA 95.38 USD
101792-742 101792-744
2-Chloro-2,2-difluoroethanol ≥96%
2-Chloro-2,2-difluoroethanol

Hazard: Irritant/Refrigerate

Formula: C₂H₃ClF₂O
Boiling Pt: 93…95 °C
MDL Number: MFCD03452785
CAS Number: 464-00-6

Order Now

Specification Test Results

Boiling point 93-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR