4-[(2-Methoxyethyl)amino]-1-methylpiperidine

Supplier: Matrix Scientific

007352-1G 007352-5G
101792-530EA 44.62 USD
101792-530 101792-532
4-[(2-Methoxyethyl)amino]-1-methylpiperidine
4-[(2-Methoxyethyl)amino]-1-methylpiperidine

Hazard: Irritant

Formula: C₉H₂₀N₂O
Boiling Pt: 62…64 °C (0.03 mmHg)
MDL Number: MFCD01826433
CAS Number: 416887-38-2

Order Now

Specification Test Results

Boiling point 62-64°C/0.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR