4-(Dimethylamino)benzylamine

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)-N,N-dimethylaniline

007349-1G 007349-5G
101792-522EA 364.62 USD
101792-522 101792-524
4-(Dimethylamino)benzylamine
4-(Dimethylamino)benzylamine

Hazard: Corrosive/Stored under Argon/Air Sensitive

Formula: C₉H₁₄N₂
Boiling Pt: 82…84 °C (0.8 mmHg)
MDL Number: MFCD00466364
CAS Number: 19293-58-4

Order Now

Specification Test Results

Boiling point 150°C/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR