Bis(1-methylpiperidin-4-yl)amine

Supplier: Matrix Scientific

007345-250MG 007345-1G
101792-514EA 126.15 USD
101792-514 101792-516
Bis(1-methylpiperidin-4-yl)amine
Bis(1-methylpiperidin-4-yl)amine

Hazard: Irritant

Formula: C₁₂H₂₅N₃
Boiling Pt: 102…105 °C (0.6 mmHg)
MDL Number: MFCD03426080
CAS Number: 117927-28-3

Order Now

Specification Test Results

Boiling point 102-105°C/0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR