1-Acetyl-4-[(2-methoxyethyl)amino]piperidine

Supplier: Matrix Scientific

Synonyms: 1-Acetyl-N-(methoxyethyl)piperidin-4-amine

007342-1G 007342-5G
101792-500EA 64.62 USD
101792-500 101792-502
1-Acetyl-4-[(2-methoxyethyl)amino]piperidine
1-Acetyl-4-[(2-methoxyethyl)amino]piperidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 168…171 °C (1 mmHg)
MDL Number: MFCD03452783
CAS Number: 849021-41-6

Order Now

Specification Test Results

Boiling point 168-171°C/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR