2,3-Dihydrobenzo[b]furan-5-carbaldehyde ≥98%

Supplier: Matrix Scientific

007319-5G 007319-25G
101792-438EA 29.23 USD
101792-438 101792-440
2,3-Dihydrobenzo[b]furan-5-carbaldehyde ≥98%
2,3-Dihydrobenzo[b]furan-5-carbaldehyde

Hazard: Irritant

Formula: C₉H₈O₂
Boiling Pt: 84 °C (0.1 mmHg)
MDL Number: MFCD00068058
CAS Number: 55745-70-5

Order Now

Specification Test Results

Boiling point 84°C/0.1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR