4-Amino-1-methylpiperidine dihydrochloride ≥95%
Supplier: Matrix Scientific
007267-5G
007267-25G
101792-290EA
35.38
USD
101792-290
101792-292
4-Amino-1-methylpiperidine dihydrochloride ≥95%
4-Amino-1-methylpiperidine dihydrochloride
Hazard: Irritant
Formula:
C₆H₁₆Cl₂N₂ MW: 187.11 g/mol Melting Pt: 247 °C |
CAS Number:
1193-03-9 |
Specification Test Results
Boiling point | 57-60°C/12m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...