1,1-Dibromo-2,2,2-trifluoroethane ≥99%
Supplier: Matrix Scientific
007195-5G
007195-25G
101792-162EA
103.08
USD
101792-162
101792-164
1,1-Dibromo-2,2,2-trifluoroethane ≥99%
1,1-Dibromo-2,2,2-trifluoroethane
Hazard: Ozone Depleter
Formula:
Br₂HCCF₃ Boiling Pt: 73.5-74.5°C |
CAS Number:
354-30-3 |
Specification Test Results
Boiling point | 73.5-74.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...