(S)-(-)-1,1,1-Trifluorononan-2-ol ≥ ee 98%

Supplier: Matrix Scientific

007101-1G 007101-5G
101792-024EA 118.46 USD
101792-024 101792-026
(S)-(-)-1,1,1-Trifluorononan-2-ol ≥ ee 98%
(S)-(-)-1,1,1-Trifluorononan-2-ol

Hazard: Irritant

Formula: C₉H₁₇F₃O
Boiling Pt: 67…68 °C (6 mmHg)
MDL Number: MFCD06799353
CAS Number: 147991-84-2

Order Now

Specification Test Results

Boiling point 67-68°C/6mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR