1,2,2-Trifluoro-2-hydroxy-1-(trifluoromethyl)ethanesulfonic acid sultone ≥98%

Supplier: Matrix Scientific

Synonyms: 3,4,4-Trifluoro-3-(trifluoromethyl)-1,2-oxathietane 2,2-dioxide

007091-1G 007091-5G
101792-004EA 100 USD
101792-004 101792-006
1,2,2-Trifluoro-2-hydroxy-1-(trifluoromethyl)ethanesulfonic acid sultone ≥98%
1,2,2-Trifluoro-2-hydroxy-1-(trifluoromethyl)ethanesulfonic acid sultone

Hazard: Corrosive/Store in Freezer

Formula: C₃F₆O₃S
MW: 230.09 g/mol
Boiling Pt: 45.6°C
MDL Number: MFCD00082475
CAS Number: 773-15-9

Order Now

Specification Test Results

Boiling point 45.6°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR