N-Methyl-1-naphthylmethylamine ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Methylaminomethyl)naphthalene

007083-5G 007083-25G
101791-986EA 26.15 USD
101791-986 102663-566
N-Methyl-1-naphthylmethylamine ≥98%
N-Methyl-1-naphthylmethylamine

Hazard: Irritant

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 104…105 °C (0.05 mmHg)
MDL Number: MFCD00144934
CAS Number: 14489-75-9

Order Now

Specification Test Results

Boiling point 104-105°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR