4-Cyclopropylamino-1-methylpiperidine

Supplier: Matrix Scientific

006987-1G 006987-5G
101791-746EA 183.08 USD
101791-746 101791-748
4-Cyclopropylamino-1-methylpiperidine
4-Cyclopropylamino-1-methylpiperidine

Hazard: Irritant

Formula: C₉H₁₈N₂
Boiling Pt: 36°C/0.05mm
MDL Number: MFCD01941329
CAS Number: 387358-48-7

Order Now

Specification Test Results

Boiling point 36°C/0.05mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR