1-Benzyl-4-cyclopropylaminopiperidine

Supplier: Matrix Scientific

006983-1G 006983-5G
101791-730EA 58.46 USD
101791-730 101791-732
1-Benzyl-4-cyclopropylaminopiperidine
1-Benzyl-4-cyclopropylaminopiperidine

Hazard: Irritant

Formula: C₁₅H₂₂N₂
Boiling Pt: 118°C/0.1mm
MDL Number: MFCD01308091
CAS Number: 387358-47-6

Order Now

Specification Test Results

Boiling point 118°C/0.1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR