1-Acetyl-4-cyclopropylaminopiperidine
Supplier: Matrix Scientific
Synonyms:
1-[4-(Cyclopropylamino)piperidin-1-yl]ethan-1-one
006982-1G
006982-5G
101791-726EA
53.85
USD
101791-726
101791-728
1-Acetyl-4-cyclopropylaminopiperidine
1-Acetyl-4-cyclopropylaminopiperidine
Hazard: Irritant
Formula:
C₁₀H₁₈N₂O MW: 182.27 g/mol Boiling Pt: 130 °C (0.15 mmHg) |
MDL Number:
MFCD01941331 CAS Number: 387358-46-5 |
Specification Test Results
Boiling point | 130°C/0.15m |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...