1-n-Propyl-4-cyclopropylaminopiperidine

Supplier: Matrix Scientific

Synonyms: N-Cyclopropyl-1-propylpiperidin-4-amine

006981-1G 006981-5G
101791-722EA 44.62 USD
101791-722 101791-724
1-n-Propyl-4-cyclopropylaminopiperidine
1-n-Propyl-4-cyclopropylaminopiperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂
MW: 182.31 g/mol
Boiling Pt: 80 °C (2 mmHg)
MDL Number: MFCD01941332
CAS Number: 387358-45-4

Order Now

Specification Test Results

Boiling point 80°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR