Ethyl picolinoylacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-oxo-3-(2-pyridyl)propionate, Ethyl 3-oxo-3-(pyridin-2-yl)propanoate

006954-1G 006954-5G
101791-662EA 52.31 USD
101791-662 101791-664
Ethyl picolinoylacetate ≥95%
Ethyl picolinoylacetate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Boiling Pt: 100…104 °C (0.3 mmHg)
MDL Number: MFCD00094022
CAS Number: 26510-52-1

Order Now

Specification Test Results

Boiling point 100-104°C/0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR