[6-(Trifluoromethyl)pyridin-3-yl]methylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Aminomethyl)-6-(trifluoromethyl)pyridine

006912-1G 006912-5G
101791-546EA 98.46 USD
101791-546 101791-548
[6-(Trifluoromethyl)pyridin-3-yl]methylamine ≥95%
[6-(Trifluoromethyl)pyridin-3-yl]methylamine

Hazard: Irritant

Formula: C₇H₇F₃N₂·HCl
MW: 176.14 g/mol
Boiling Pt: 62…64 °C (0.4 mmHg)
MDL Number: MFCD01862649
CAS Number: 387350-39-2

Order Now

Specification Test Results

Boiling point 62-64°C/0.4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR