N,N-Diethyl-2,3,3,3-tetrafluoropropionamide ≥97%
Supplier: Matrix Scientific
Synonyms:
N,N-Diethyl-2,3,3,3-tetrafluoropropanamide
006611-5G
006611-25G
101790-814EA
127.69
USD
101790-814
101790-816
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide ≥97%
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide
Hazard: Irritant
Formula:
C₇H₁₁F₄NO MW: 201.17 g/mol Boiling Pt: 58°C/5mm |
MDL Number:
MFCD00069160 CAS Number: 392-63-2 |
Specification Test Results
Boiling point | 58°C/5mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...