Ethyl-4-(trifluoromethyl)benzoate ≥97%

Supplier: Matrix Scientific

006388-5G 006388-25G 006388-100G
101790-486EA 30.77 USD
101790-486 101790-488 101790-490
Ethyl-4-(trifluoromethyl)benzoate ≥97%
Ethyl-4-(trifluoromethyl)benzoate

Hazard: Irritant

Formula: C₁₀H₉F₃O₂
Boiling Pt: 81 °C (5 mmHg)
MDL Number: MFCD00013563
CAS Number: 583-02-8

Order Now

Specification Test Results

Boiling point 81°C/5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR