Ethyl-2-bromo-2,3,3,3-tetrafluoropropionate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl perfluoro-2-bromopropanoate

006041-1G 006041-5G
101789-960EA 147.69 USD
101789-960 101789-962
Ethyl-2-bromo-2,3,3,3-tetrafluoropropionate ≥97%
Ethyl-2-bromo-2,3,3,3-tetrafluoropropionate

Hazard: Irritant

Formula: C₅H₅BrF₄O₂
MW: 253 g/mol
Boiling Pt: 122-124°C
MDL Number: MFCD00153721
CAS Number: 10186-73-9

Order Now

Specification Test Results

Boiling point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR