4-(Difluoromethoxy)iodobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Difluoromethoxy)-4-iodobenzene

006022-5G 006022-25G
101789-934EA 29.23 USD
101789-934 101789-936
4-(Difluoromethoxy)iodobenzene ≥98%
4-(Difluoromethoxy)iodobenzene

Hazard: Irritant/Refrigerate

Formula: C₇H₅F₂IO
MW: 270.0153 g/mol
Boiling Pt: 86-88°C/6mm
MDL Number: MFCD00085008
CAS Number: 128140-82-9

Order Now

Specification Test Results

Boiling point 86-88°C/6mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR