4-(Difluoromethoxy)toluene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Difluoromethoxy)-4-methylbenzene

006016-1G 006016-5G
101789-918EA 95.38 USD
101789-918 101789-920
4-(Difluoromethoxy)toluene ≥97%
4-(Difluoromethoxy)toluene

Hazard: Irritant

Formula: C₈H₈F₂O
MW: 158.15 g/mol
Boiling Pt: 50…52 °C (15 mmHg)
MDL Number: MFCD00042253
CAS Number: 1583-83-1

Order Now

Specification Test Results

Boiling point 50-52°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR