1,1,1,3,3-Pentachloro-3-fluoroacetone

Supplier: Matrix Scientific

Synonyms: Pentachlorofluoroacetone

005983-1G 005983-5G
101789-880EA 73.85 USD
101789-880 101789-882
1,1,1,3,3-Pentachloro-3-fluoroacetone
1,1,1,3,3-Pentachloro-3-fluoroacetone

TCA listed: T
Hazard: Irritant-Harmful

Formula: C₃Cl₅FO
MW: 248.29 g/mol
Boiling Pt: 163…166 °C
CAS Number: 2378-08-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR