Pentafluoroethyl phenyl ketone ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2,3,3,3-Pentafluoropropiophenone, Pentafluoroethyl phenyl ketone
005981-1G
005981-5G
101789-872EA
176.92
USD
101789-872
101789-874
Pentafluoroethyl phenyl ketone ≥97%
Pentafluoroethyl phenyl ketone
Hazard: Irritant
Formula:
C₉H₅F₅O MW: 224.13 g/mol Boiling Pt: 160…161 °C |
MDL Number:
MFCD00018062 CAS Number: 394-52-5 |
Specification Test Results
Boiling point | 160-161°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...