Pentafluoro(2,2,2-trifluoroethoxy)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: (2,2,2-Trifluoroethoxy)pentafluorobenzene

005890-5G 005890-25G
101789-706EA 76.92 USD
101789-706 101789-708
Pentafluoro(2,2,2-trifluoroethoxy)benzene ≥97%
Pentafluoro(2,2,2-trifluoroethoxy)benzene

Hazard: Irritant

Formula: C₈H₂F₈O
MW: 266.09 g/mol
Boiling Pt: 146-147°C
MDL Number: MFCD00155915
CAS Number: 6669-03-0

Order Now

Specification Test Results

Boiling point 146-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR